Search by lot number or partial lot number
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 60mL (2 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 60 mL |
Closure Color | White |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container bagged separately, non-sterile, no label, white caps |
Diameter (Metric) | 48 mm |
Label | No Label |
No. per Case | 500/Cs. |
No. per Pack | 500/Pk. |
Unit Size | Case of 500 |
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Sterile |
Capacity (Metric) | 60 mL |
Closure Color | White |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container assembled, sterile, tabbed label, white caps |
Diameter (Metric) | 48 mm |
Label | Tabbed |
No. per Case | 500/Cs. |
No. per Pack | 100/Pk. |
Unit Size | Case of 500 |
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Sterility | Sterile |
Capacity (Metric) | 60 mL |
Closure Color | Orange |
Closure Size | 48 mm |
Closure Type | Bio-Tite |
Description | Cap and container assembled, sterile, tabbed label, orange caps |
Diameter (Metric) | 48 mm |
Label | Tabbed |
Unit Size | Case of 500 |
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 60 mL |
Closure Color | Yellow |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container assembled, non-sterile, no label, yellow caps |
Diameter (Metric) | 48 mm |
Label | No Label |
No. per Case | 500/Cs. |
No. per Pack | 100/Pk. |
Unit Size | Case of 500 |
Product Type | Specimen Container |
Closure Included | Yes |
Material | Virgin Polypropylene |
Shape | Cylindrical |
Sterility | Non-sterile |
Capacity (Metric) | 60 mL |
Closure Color | Orange |
Closure Size | 48 mm |
Closure Type | Bio-Tite Lid |
Description | Cap and container bagged separately, non-sterile, no label, orange caps |
Diameter (Metric) | 48 mm |
Label | No Label |
No. per Case | 500/Cs. |
No. per Pack | 500/Pk. |
Unit Size | Case of 500 |
Catalog Number | Specifications | Unit Size | Closure Color | Label | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
030181 also known as 03 0181 | Case of 500 | White | No Label | Cap and container bagged separately, non-sterile, no label, white caps | 199.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
030007 also known as 03 0007 | Case of 500 | White | Tabbed | Cap and container assembled, sterile, tabbed label, white caps | 224.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
030006 also known as 03 0006 | Case of 500 | Orange | Tabbed | Cap and container assembled, sterile, tabbed label, orange caps | 224.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
030193 also known as 03 0193 | Case of 500 | Yellow | No Label | Cap and container assembled, non-sterile, no label, yellow caps | 199.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
030195 also known as 03 0195 | Case of 500 | Orange | No Label | Cap and container bagged separately, non-sterile, no label, orange caps | 199.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Available Options: