Thermo Scientific Chemicals

Diethyl methylphosphonite, Thermo Scientific Chemicals

Catalog number: 030357.02
0.5 g, Each
Thermo Scientific Chemicals

Diethyl methylphosphonite, Thermo Scientific Chemicals

Catalog number: 030357.02
0.5 g, Each
Quantity
Catalog number: 030357.02
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15715-41-0
IUPAC Name
diethyl methylphosphonite
Molecular Formula
C5H13O2P
InChI Key
NSSMTQDEWVTEKN-UHFFFAOYSA-N
SMILES
CCOP(C)OCC
Appearance (Color)
Colorless to yellow
Form
Liquid
Assay (GC)
≥94%
Infrared spectrum
Conforms
Refractive Index
1.4168

Description

Dimethyl methylphosphonate is a phosphonite reagent used in the generation of phosphinic acid derivative via Arbuzov reaction and as a cyclization reagentin the preparation of 2-substituted penems. It is a chiral intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text