Specifications
Chemical Name or MaterialTriethyl 2-phosphonopentanoate
CAS35051-49-1
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more