Triethyl 2-phosphonopentanoate, 98%, Thermo Scientific Chemicals
Triethyl 2-phosphonopentanoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl 2-phosphonopentanoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 030413.09
also known as 030413-09
Price (USD)/ Each
482.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
482.00
Add to cart
Triethyl 2-phosphonopentanoate, 98%, Thermo Scientific Chemicals
Catalog number030413.09
Price (USD)/ Each
482.00
-
Add to cart
Specifications
Chemical Name or MaterialTriethyl 2-phosphonopentanoate
CAS35051-49-1
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only