Thermo Scientific Chemicals

Zirconium silicate, Thermo Scientific Chemicals

Catalog number: 030477.36
500 g, Each
Thermo Scientific Chemicals

Zirconium silicate, Thermo Scientific Chemicals

Catalog number: 030477.36
500 g, Each
Quantity
Catalog number: 030477.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10101-52-7
IUPAC Name
zirconium(4+) silicate
Molecular Formula
O4SiZr
InChI Key
GFQYVLUOOAAOGM-UHFFFAOYSA-N
SMILES
[Zr+4].[O-][Si]([O-])([O-])[O-]
Appearance (Color)
White to pale cream
Form
Powder

Description

Zirconium silicate is used for manufacturing refractory materials for applications where resistance to corrosion by alkali materials is required. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. It can be also present in some cements. Another use of zirconium silicate is as beads for milling and grinding. Thin films of zirconium silicate and hafnium silicate produced by chemical vapor deposition. Potential alternative gate dielectric to silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text