Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 030482.22
100 g, Each
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 030482.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 030482.22
also known as 030482-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13477-34-4
IUPAC Name
calcium bis(nitric acid) tetrahydrate
Molecular Formula
CaH10N2O10
InChI Key
DKCJOKYHYIAZQH-UHFFFAOYSA-N
SMILES
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
Total Metal Impurities
0.02% max.

Description

Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text