Thermo Scientific Chemicals

Iron(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 030513.14
25 g, Each
Thermo Scientific Chemicals

Iron(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 030513.14
25 g, Each
Quantity
Catalog number: 030513.14
also known as 030513-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1345-25-1
IUPAC Name
oxoiron
Molecular Formula
FeO
InChI Key
UQSXHKLRYXJYBZ-UHFFFAOYSA-N
SMILES
O=[Fe]
Total Metal Impurities
0.5% max.

Description

In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalystIron(II) oxide is used in manufacturing of green, heat-absorbing glass; in steel manufacture; and as a catalyst. It is used as a dye or pigment in pottery, glazes and glasses. It finds application as heat-absorbing glass used in buildings and automobiles. It is used as a precursor in the production of steel. It is also used in cosmetics and some tattoo inks. It can also be used for filtering phosphates from home aquaria and acts as a catalyst in a number of industrial and chemical operations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text