Thermo Scientific Chemicals

Manganese(III) acetate hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 030519.09
10 g, Each
Thermo Scientific Chemicals

Manganese(III) acetate hydrate, 96%, Thermo Scientific Chemicals

Catalog number: 030519.09
10 g, Each
Quantity
Catalog number: 030519.09
also known as 030519-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19513-05-4
IUPAC Name
manganese(3+) triacetate
Molecular Formula
C6H9MnO6
InChI Key
AHSBSUVHXDIAEY-UHFFFAOYSA-K
SMILES
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Assay (unspecified)
>= 95% (≥15% Mn)
Appearance (Color)
Orange to Brown
Form
solid

Description

It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text