Thermo Scientific Chemicals

Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals

Catalog number: 030529.30
250 g, Each
Thermo Scientific Chemicals

Lithium 2,4-pentanedionate, 99.5%, Thermo Scientific Chemicals

Catalog number: 030529.30
250 g, Each
Quantity
Catalog number: 030529.30
also known as 030529-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18115-70-3
IUPAC Name
lithium(1+) (2Z)-4-oxopent-2-en-2-olate
Molecular Formula
C5H7LiO2
InChI Key
JTEOOCAGEXVCBQ-LNKPDPKZSA-M
SMILES
[Li+].C\C([O-])=C\C(C)=O
Form
Powder or crystalline powder or crystals or granules
Appearance (Color)
White to pale yellow
Assay from Suppliers CofA
≥97.5%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text