Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals
Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 030532.18
also known as 030532-18
Price (USD)/ Each
454.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
454.00
Add to cart
Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals
Catalog number030532.18
Price (USD)/ Each
454.00
-
Add to cart
Specifications
Chemical Name or MaterialTin(IV) chloride bis(2,4-pentanedionate)
Melting Point199°C
CAS16919-46-3
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only