1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals

Oxidation, reduction indicator
Have Questions?
Quantity:
10 g
Catalog number 030557.09
also known as 030557-09
Price (USD)/ Each
206.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
206.00
Add to cart
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Catalog number030557.09
Price (USD)/ Each
206.00
-
Add to cart
Specifications
Chemical Name or Material1,10-Phenanthroline iron(II) perchlorate
CAS14586-54-0
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
1,10-Phenanthroline iron(II) perchlorate is used as an oxidation, reduction indicator. It is used for the detection, identification, analysis, of chemical, biological, or pathologic processes or conditions. Agents that are capable of inserting themselves between the successive bases in DNA, thus kinking, uncoiling or otherwise deforming it and therefore preventing its proper functioning. They are used in the study of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only