Thermo Scientific Chemicals

Linear alkylbenzenesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 030574.D5
3 x 1 kg, Each
Thermo Scientific Chemicals

Linear alkylbenzenesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: 030574.D5
3 x 1 kg, Each
Quantity
Catalog number: 030574.D5
also known as 030574-D5
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68584-22-5
IUPAC Name
4-(dodecan-3-yl)benzene-1-sulfonic acid
Molecular Formula
C18H30O3S
InChI Key
QJRVOJKLQNSNDB-UHFFFAOYNA-N
SMILES
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Clear dark yellow to dark orange
Form
Viscous liquid
Assay (unspecified)
≥96.0%

Description

Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text