Thermo Scientific Chemicals

Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 030638.09
10 g, Each
Thermo Scientific Chemicals

Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals

Catalog number: 030638.09
10 g, Each
Quantity
Catalog number: 030638.09
also known as 030638-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62-55-5
IUPAC Name
ethanethioamide
Molecular Formula
C2H5NS
InChI Key
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
SMILES
CC(N)=S
Residue on ignition
0.05%
Melting Point
111-114?
Assay (Titration)
>/+ 99.0
Comment
Clarity of a 2% solution P.T.

Description

Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text