Thermo Scientific Chemicals

Acetone, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 030698.K2
1 L, Each
Thermo Scientific Chemicals

Acetone, ACS, 99.5+%, Thermo Scientific Chemicals

Catalog number: 030698.K2
1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 030698.K2
also known as 030698-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-64-1
IUPAC Name
propan-2-one
Molecular Formula
C3H6O
InChI Key
CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES
CC(C)=O
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥99.5% (corrected for water)
Free acid (titration)
≤0.0003meq/g
Free base (titration)
≤0.0006meq/g

Description

Acetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glasswares due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon, photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text