Thermo Scientific Chemicals

Aluminum iodide, 95%, Thermo Scientific Chemicals

Catalog number: 030714.14
25 g, Each
Thermo Scientific Chemicals

Aluminum iodide, 95%, Thermo Scientific Chemicals

Catalog number: 030714.14
25 g, Each
Quantity
Catalog number: 030714.14
also known as 030714-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-23-8
IUPAC Name
aluminium(3+) triiodide
Molecular Formula
AlI3
InChI Key
CECABOMBVQNBEC-UHFFFAOYSA-K
SMILES
[Al+3].[I-].[I-].[I-]
Assay (unspecified)
94.1% min

Description

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text