Aluminum iodide, 95%, Thermo Scientific Chemicals
Aluminum iodide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum iodide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 030714.22
also known as 030714-22
Price (USD)/ Each
311.65
Online exclusive
346.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
311.65
Online exclusive
346.00 
Save 34.35 (10%)
Add to cart
Aluminum iodide, 95%, Thermo Scientific Chemicals
Catalog number030714.22
Price (USD)/ Each
311.65
Online exclusive
346.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7784-23-8
IUPAC Namealuminium(3+) triiodide
Molecular FormulaAlI3
InChI KeyCECABOMBVQNBEC-UHFFFAOYSA-K
SMILES[Al+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only