Thermo Scientific Chemicals

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%, Thermo Scientific Chemicals

Catalog number: 030715.30
250 g, Each
Thermo Scientific Chemicals

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%, Thermo Scientific Chemicals

Catalog number: 030715.30
250 g, Each
Quantity
Catalog number: 030715.30
also known as 030715-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32823-06-6
Assay (unspecified)
Al₂O₃ 19.4% min
Assay (unspecified)
P₂O₅ 78.9% min

Description

Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text