Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%
Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%
Thermo Scientific Chemicals

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%

CAS: 32823-06-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 x 1 kg
Catalog number 030715.D9
also known as 030715-D9
Price (USD)
374.00
Each
-
Add to cart
Quantity:
5 x 1 kg
Request bulk or custom format
Price (USD)
374.00
Each
Add to cart
Chemical Identifiers
CAS32823-06-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Al₂O₃ 19.4% min
Assay (unspecified)P₂O₅ 78.9% min
Ask our AI about this Product
Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link