Thermo Scientific Chemicals

Ammonium benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 030753.22
100 g, Each
Thermo Scientific Chemicals

Ammonium benzoate, 99%, Thermo Scientific Chemicals

Catalog number: 030753.22
100 g, Each
Quantity
Catalog number: 030753.22
also known as 030753-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1863-63-4
IUPAC Name
ammonium benzoate
Molecular Formula
C7H9NO2
InChI Key
VWSRWGFGAAKTQG-UHFFFAOYSA-N
SMILES
[NH4+].[O-]C(=O)C1=CC=CC=C1
Assay (Titration)
> 98.5%
Form
Crystalline powder

Description

Ammonium benzoate is used predominantly in industries which acts as a corrosion inhibitors and anti-scaling agents . It is also used as paint additives and coating additives. Other consumer uses include cleaning and furnishing Care products, paints and coatings, personal care products, water treatment products. Ammonium benzoate is also used as chemical analysis reagents, in electronics industry, used for the production of electrolytic capacitors and also used in pharmaceutical industries. Used as adhesives, metal rust treatment and rubber additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text