Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

For Synthesis and Liquid Chromatography
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
4 x 1 L
4 L
Catalog number 030760.K2
also known as 030760-K2
Price (USD)/ Each
59.65
Online exclusive
65.60 
Save 5.95 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
65.60 
Save 5.95 (9%)
Add to cart
Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals
Catalog number030760.K2
Price (USD)/ Each
59.65
Online exclusive
65.60 
Save 5.95 (9%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Color (APHA)20, max
Water Content (Karl Fischer Titration)0.05%, max
Assay (GC)≥99.0%
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities: Color (APHA) 20, Peroxide (as H2O2) 0.015%, Residue after evaporation 0.03%, H2O 0.05%
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only