Thermo Scientific Chemicals

(±)-Propylene oxide, >99%, Thermo Scientific Chemicals

Catalog number: 030765.K2
1 L, Each
Thermo Scientific Chemicals

(±)-Propylene oxide, >99%, Thermo Scientific Chemicals

Catalog number: 030765.K2
1 L, Each
Quantity
Catalog number: 030765.K2
also known as 030765-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-56-9
IUPAC Name
2-methyloxirane
Molecular Formula
C3H6O
InChI Key
GOOHAUXETOMSMM-UHFFFAOYNA-N
SMILES
CC1CO1
Appearance (Color)
Colorless
Form
Liquid
Assay (GC)
≥99.0%
Refractive Index
1.3640-1.3680 @ 20°C

Description

Propylene oxide is used as a precursor in the production of polyether polyols, which is utilized for making polyurethane plastics, polypropylene glycols, propylene glycol ethers and propylene carbonate. It is involved in the preparation of biological samples for electron microscopy. It plays a vital role in the removal of residual ethanol previously used for dehydration. Further, it is used in thermobaric weapons, and microbial fumigation. It is also used as solvents, and in the preparation of surfactants, lubricants and in oil demulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text