Thermo Scientific Chemicals

Salicylic acid, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 030782.0E
2500 g, Each
Thermo Scientific Chemicals

Salicylic acid, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 030782.0E
2500 g, Each
Quantity
Catalog number: 030782.0E
also known as 030782-0E
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
69-72-7
IUPAC Name
2-hydroxybenzoic acid
Molecular Formula
C7H6O3
InChI Key
YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1O
Assay (HPLC)
> 99.0% C7H6O3
Residue on ignition
0.01%
Impurity content
Cl 0.001%
Impurity content
Fe 2ppm
Impurity content
SO₄ 0.003%

Description

Salicylic acid is used as a food preservative as well as used in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in the dye industry for making azo direct dyes and acidic mordant dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 1
    Reproductive Toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H318 - Causes serious eye damage

    H361d - Suspected of damaging the unborn child

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician