n-Tetradecane, 99+%, Thermo Scientific Chemicals
n-Tetradecane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Tetradecane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 030809.36
also known as 030809-36
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
n-Tetradecane, 99+%, Thermo Scientific Chemicals
Catalog number030809.36
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS629-59-4
IUPAC Nametetradecane
Molecular FormulaC14H30
InChI KeyBGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay (unspecified)Hydrocarbons with Carbon number, C, > 14: 0.4% mac by GC
Assay (unspecified)Hydrocarbons with Carbon number, C, < 14: 0.4% max ( by GC)
Assay (GC)> 99.0%
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only