Specifications
Chemical Name or MaterialTetra-n-hexylammonium perchlorate
CAS4656-81-9
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more