Tetramethylammonium perchlorate, Thermo Scientific Chemicals
Tetramethylammonium perchlorate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylammonium perchlorate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 030834.14
also known as 030834-14
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Tetramethylammonium perchlorate, Thermo Scientific Chemicals
Catalog number030834.14
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2537-36-2
IUPAC Nametetramethylazanium perchlorate
Molecular FormulaC4H12ClNO4
InChI KeyZCWKIFAQRXNZCH-UHFFFAOYSA-M
SMILESC[N+](C)(C)C.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥97.0% (after drying)
Assay (special)Halogens (as Cl): ≤0.01%
Water Content≤7.0%
Tetramethylammonium perchlorate acts as an intermediate in organic and chemical synthesis. It is used as a mobile phase in high performance liquid chromatographic analysis and in the quantitation of olanzapine clozapine and N-desmethylclozapine. It acts as a supporting electrolyte in the electrochemical reduction of electrochemical reduction of 1-bromodecanr, 1-iododecane,2-iodooctane and 2-bromooctane at mercury cathodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only