Thermo Scientific Chemicals

Tetramethylammonium perchlorate, Thermo Scientific Chemicals

Catalog number: 030834.22
100 g, Each
Thermo Scientific Chemicals

Tetramethylammonium perchlorate, Thermo Scientific Chemicals

Catalog number: 030834.22
100 g, Each
Quantity
Catalog number: 030834.22
also known as 030834-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2537-36-2
IUPAC Name
tetramethylazanium perchlorate
Molecular Formula
C4H12ClNO4
InChI Key
ZCWKIFAQRXNZCH-UHFFFAOYSA-M
SMILES
C[N+](C)(C)C.[O-][Cl](=O)(=O)=O
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥97.0% (after drying)
Assay (special)
Halogens (as Cl): ≤0.01%
Water Content
≤7.0%

Description

Tetramethylammonium perchlorate acts as an intermediate in organic and chemical synthesis. It is used as a mobile phase in high performance liquid chromatographic analysis and in the quantitation of olanzapine clozapine and N-desmethylclozapine. It acts as a supporting electrolyte in the electrochemical reduction of electrochemical reduction of 1-bromodecanr, 1-iododecane,2-iodooctane and 2-bromooctane at mercury cathodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text