Thermo Scientific Chemicals
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals
Catalog number: 030854.03
1 g, Each
Thermo Scientific Chemicals
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals
Catalog number: 030854.03
1 g, Each
Quantity
Catalog number: 030854.03
also known as 030854-03
Price (USD)
Quantity
-
Have Questions?
Description
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane is used as a macrocyclic chelating agent in coordination chemistry. It is also used as a ligand which bind strongly to a wide range of metal ions and as crown ethers.