Thermo Scientific Chemicals

1-Pentanol, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 030898.K7
4 L, Each
Thermo Scientific Chemicals

1-Pentanol, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 030898.K7
4 L, Each
Quantity
Catalog number: 030898.K7
also known as 030898-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-41-0
IUPAC Name
pentan-1-ol
Molecular Formula
C5H12O
InChI Key
AMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILES
CCCCCO
Residual water
≤0.5%
Impurity content
Acids and esters: ≤0.075meq/g
Residue on Evaporation
≤0.003%
Color (APHA)
≤30
Impurity content
Carbonyl (as HCHO): ≤0.1%

Description

1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text