Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals

Catalog number: 030910.18
50 g, Each
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals

Catalog number: 030910.18
50 g, Each
Quantity
Catalog number: 030910.18
also known as 030910-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5144-89-8
IUPAC Name
1,10-phenanthroline hydrate
Molecular Formula
C12H10N2O
InChI Key
PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Comment
Suitability for determining iron P.T.
Comment
Suitability as redox indicator P.T.

Description

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text