Thermo Scientific Chemicals

Pentacarbonyliron, 99.5%, Thermo Scientific Chemicals

Catalog number: 031113.14
25 g, Each
Thermo Scientific Chemicals

Pentacarbonyliron, 99.5%, Thermo Scientific Chemicals

Catalog number: 031113.14
25 g, Each
Quantity
Catalog number: 031113.14
also known as 031113-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13463-40-6
IUPAC Name
pentakis(methanidylidyneoxidanium) iron
Molecular Formula
C5FeO5
InChI Key
FYOFOKCECDGJBF-UHFFFAOYSA-N
SMILES
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Comment
Material may have to be filtered through glass wool before use.
Comment
May contain particulate matter.
Form
Liquid
Appearance (Color)
Yellow-orange to brown
Comment
Keep residue moist, may be pyrophoric if allowed to dry.

Description

Pentacarbonyliron is used as a catalyst in organic reactions, and as carbonyl iron for high-frequency coils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text