Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals
Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Catalog number 031124.14
Price (USD)/ Each
70.65
Online exclusive
78.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
78.50 
Save 7.85 (10%)
Add to cart
Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals
Catalog number031124.14
Price (USD)/ Each
70.65
Online exclusive
78.50 
Save 7.85 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPotassium trioxalatoferrate(III) trihydrate
CAS5936-11-8
ColorGreen
Health Hazard 1H302+H312
Health Hazard 2GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
View more
Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only