Thermo Scientific Chemicals

Iron(II) acetate, anhydrous, Fe 29.5% min, Thermo Scientific Chemicals

Catalog number: 031140.22
100 g, Each
Thermo Scientific Chemicals

Iron(II) acetate, anhydrous, Fe 29.5% min, Thermo Scientific Chemicals

Catalog number: 031140.22
100 g, Each
Quantity
Catalog number: 031140.22
also known as 031140-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3094-87-9
IUPAC Name
λ²-iron(2+) diacetate
Molecular Formula
C4H6FeO4
InChI Key
LNOZJRCUHSPCDZ-UHFFFAOYSA-L
SMILES
[Fe++].CC([O-])=O.CC([O-])=O
Assay (unspecified)
91% min

Description

Iron(II) acetate is used as a mordant by the dye industry, as a catalyst in organic oxidation reactions and in carbon nanotube synthesis. It is used as a precursor in the production of other iron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text