Thermo Scientific Chemicals

Iron(II) bromide, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog number: 031154.06
5 g, Each
Thermo Scientific Chemicals

Iron(II) bromide, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog number: 031154.06
5 g, Each
Quantity
Catalog number: 031154.06
also known as 031154-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-46-0
IUPAC Name
λ²-iron(2+) dibromide
Molecular Formula
Br2Fe
InChI Key
GYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES
[Fe++].[Br-].[Br-]
Assay (unspecified)
97.6% min

Description

Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text