5-Chloro-1,10-phenanthroline, Thermo Scientific Chemicals
5-Chloro-1,10-phenanthroline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-1,10-phenanthroline, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
Catalog number 031180.04
also known as 031180-04
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
Add to cart
5-Chloro-1,10-phenanthroline, Thermo Scientific Chemicals
Catalog number031180.04
Price (USD)/ Each
210.65
Online exclusive
234.00 
Save 23.35 (10%)
-
Add to cart
Specifications
Chemical Name or Material5-Chloro-1,10-phenanthroline
CAS4199-89-7
Melting Point123°C
Recommended StorageAmbient temperatures
EINECS Number224-098-1
View more
Used to prepare 1,10-phenanthrolinium cation. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only