Thermo Scientific Chemicals

n-Decane, 99+%, Thermo Scientific Chemicals

Catalog number: 031209.09
10 g, Each
Thermo Scientific Chemicals

n-Decane, 99+%, Thermo Scientific Chemicals

Catalog number: 031209.09
10 g, Each
Quantity
Catalog number: 031209.09
also known as 031209-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-18-5
IUPAC Name
decane
Molecular Formula
C10H22
InChI Key
DIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILES
CCCCCCCCCC
Appearance (Color)
Colorless [EU]
Form
Clear liquid [EU & US]
Assay (unspecified)
≤C-10 carbon: ≤0.5% [US]
Assay (GC)
≥99.0% [EU & US]
Refractive Index
1.4100-1.4130 @ 20?C [EU]

Description

n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text