Thermo Scientific Chemicals

1-Decene, 96%, remainder isomers, Thermo Scientific Chemicals

Catalog number: 031213.36
500 g, Each
Thermo Scientific Chemicals

1-Decene, 96%, remainder isomers, Thermo Scientific Chemicals

Catalog number: 031213.36
500 g, Each
Quantity
Catalog number: 031213.36
also known as 031213-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-05-9
IUPAC Name
dec-1-ene
Molecular Formula
C10H20
InChI Key
AFFLGGQVNFXPEV-UHFFFAOYSA-N
SMILES
CCCCCCCCC=C
Appearance (Color)
Clear colorless
Assay (GC)
≥95.0%
Form
Liquid

Description

1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text