Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals
Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
5 g
25 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031244.A5
also known as 031244-A5
Price (USD)/ Each
83.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
98.80 
Save 15.15 (15%)
-
Add to cart
Quantity:
3 kg
Request bulk or custom format
Price (USD)/ Each
83.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
98.80 
Save 15.15 (15%)
Add to cart
Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals
Catalog number031244.A5
Price (USD)/ Each
83.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
98.80 
Save 15.15 (15%)
-
Add to cart
Chemical Identifiers
CAS84-74-2
IUPAC Name1,2-dibutyl benzene-1,2-dicarboxylate
Molecular FormulaC16H22O4
InChI KeyDOIRQSBPFJWKBE-UHFFFAOYSA-N
SMILESCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay from Supplier's CofA≥99.0%
Acid Value≤0.2% (as phthalic acid)
FormLiquid
Dibutyl phthalate (DBP) is a common industrial chemicals used as plasticizers, softeners, adhesives or solvents used by a variety of industries. It is also used as an additive to adhesives or printing inks. Other applications include in PVC, paints, cosmetics and coatings of cars.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only