Thermo Scientific Chemicals

Dodecylbenzene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 031362.30
250 g, Each
Thermo Scientific Chemicals

Dodecylbenzene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 031362.30
250 g, Each
Quantity
Catalog number: 031362.30
also known as 031362-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-01-3
IUPAC Name
dodecylbenzene
Molecular Formula
C18H30
InChI Key
KWKXNDCHNDYVRT-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCC1=CC=CC=C1
Assay (special)
Carbon number distribution (C9%): Report
Form
Liquid
Appearance (Color)
Colorless to yellow
Molecular Weight
238-250 (average relative molecular weight)

Description

1-Phenyldodecane acts as a pseudo-stationary phase (PSP) marker in the preparation of microemulsion, 1-Phenyldodecane was degraded to phenylacetic acid by an oil-degrading bacterium. it is used to prepare an anionic detergent dodecylbenzene sodium sulfonate. It is a petrochemical-derived substitute for the tallow and coconut oil used in making soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text