Thermo Scientific Chemicals

1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals

Catalog number: 031390.22
100 g, Each
Thermo Scientific Chemicals

1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals

Catalog number: 031390.22
100 g, Each
Quantity
Catalog number: 031390.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-22-7
IUPAC Name
1,3-bis(phenylamino)urea
Molecular Formula
C13H14N4O
InChI Key
KSPIHGBHKVISFI-UHFFFAOYSA-N
SMILES
O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Comment
Sensitivity to chromate P.T.
Solution Test
in aqueous acetone P.T.
Residue on ignition
0.05%

Description

1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text