Thermo Scientific Chemicals

4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals

Catalog number: 031434.04
2 g, Each
Thermo Scientific Chemicals

4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals

Catalog number: 031434.04
2 g, Each
Quantity
Catalog number: 031434.04
also known as 031434-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3922-40-5
IUPAC Name
1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione
Molecular Formula
C12H8N2O2
InChI Key
SLIBCJURSADKPV-UHFFFAOYSA-N
SMILES
O=C1C=CNC2=C3NC=CC(=O)C3=CC=C12
Form
Powder or crystals or crystalline powder
Appearance (Color)
Brown
Purity
≥95.0%
Proton NMR
Conforms

Description

4,7-Dihydroxy-1,10-phenanthroline is used as a phenanthroline stain. Its iron(II) complex reacts rapidly with dissolved oxygen in aqueous solution, thereby it is utilized for the spectrophotometric determination of dissolved oxygen up to 20 ppm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text