4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals
4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
0.5 g
2 g
Catalog number 031434.09
also known as 031434-09
Price (USD)/ Each
642.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
642.00
Add to cart
4,7-Dihydroxy-1,10-phenanthroline, Thermo Scientific Chemicals
Catalog number031434.09
Price (USD)/ Each
642.00
-
Add to cart
Chemical Identifiers
CAS3922-40-5
IUPAC Name1,4,7,10-tetrahydro-1,10-phenanthroline-4,7-dione
Molecular FormulaC12H8N2O2
InChI KeySLIBCJURSADKPV-UHFFFAOYSA-N
SMILESO=C1C=CNC2=C3NC=CC(=O)C3=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)Brown
Purity≥95.0%
Proton NMRConforms
4,7-Dihydroxy-1,10-phenanthroline is used as a phenanthroline stain. Its iron(II) complex reacts rapidly with dissolved oxygen in aqueous solution, thereby it is utilized for the spectrophotometric determination of dissolved oxygen up to 20 ppm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only