Thermo Scientific Chemicals

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 031458.A1
1 kg, Each
Thermo Scientific Chemicals

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 031458.A1
1 kg, Each
Quantity
Catalog number: 031458.A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-40-3
IUPAC Name
dimethylamine
Molecular Formula
C2H7N
InChI Key
ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES
CNC
Appearance (Color)
Clear colorless
Assay (Aqueous acid-base Titration)
39.0 to 41.0%
Form
Liquid

Description

Dimethylamine 40% aqueous solution is used as a precursor in the preparation of nitrosodimethylamine. It is also used in paint additives, lubricants, lubricant additives and coating additives. It plays a major role in ion exchange agents and agricultural chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text