Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
3 kg
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031458.A5
Price (USD)/ Each
-
Quantity:
3 kg
Request bulk or custom format
Chemical Identifiers
CAS124-40-3
IUPAC Namedimethylamine
Molecular FormulaC2H7N
InChI KeyROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILESCNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)39.0 to 41.0%
FormLiquid

Dimethylamine 40% aqueous solution is used as a precursor in the preparation of nitrosodimethylamine. It is also used in paint additives, lubricants, lubricant additives and coating additives. It plays a major role in ion exchange agents and agricultural chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only