Thermo Scientific Chemicals

Ammonium dihydrogen arsenate, 98%, Thermo Scientific Chemicals

Catalog number: 031493.18
50 g, Each
Thermo Scientific Chemicals

Ammonium dihydrogen arsenate, 98%, Thermo Scientific Chemicals

Catalog number: 031493.18
50 g, Each
Quantity
Catalog number: 031493.18
also known as 031493-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13462-93-6
IUPAC Name
ammonium dihydrogen arsorate
Molecular Formula
AsH6NO4
InChI Key
VTWJXWNBCACAAK-UHFFFAOYSA-N
SMILES
[NH4+].O[As](O)([O-])=O
Assay (unspecified)
97.6% min

Description

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text