Arsenic(III) chloride, Reagent Grade, Thermo Scientific Chemicals
Arsenic(III) chloride, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Arsenic(III) chloride, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031499.30
also known as 031499-30
Price (USD)/ Each
-
Request bulk or custom format
Chemical Identifiers
CAS7784-34-1
IUPAC Nametrichloroarsane
Molecular FormulaAsCl3
InChI KeyOEYOHULQRFXULB-UHFFFAOYSA-N
SMILESCl[As](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only