Thermo Scientific Chemicals

2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals

Catalog number: 031733.K2
1 L, Each
Thermo Scientific Chemicals

2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals

Catalog number: 031733.K2
1 L, Each
Quantity
Catalog number: 031733.K2
also known as 031733-K2
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-86-4
IUPAC Name
2-methoxyethan-1-ol
Molecular Formula
C3H8O2
InChI Key
XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES
COCCO
Appearance (Color)
Clear
Color (APHA)
10
Assay (GC)
>/= 99.3%
Free acid (titration)
0.002meq/g
Residual water
0.1%

Description

2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text