2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals
2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 4 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031733.M6
also known as 031733-M6
Price (USD)/ Each
509.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
599.00 
Save 89.35 (15%)
-
Add to cart
Quantity:
4 x 4 L
Request bulk or custom format
Price (USD)/ Each
509.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
599.00 
Save 89.35 (15%)
Add to cart
2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals
Catalog number031733.M6
Price (USD)/ Each
509.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
599.00 
Save 89.35 (15%)
-
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
Color (APHA)10
Assay (GC)>/= 99.3%
Free acid (titration)0.002meq/g
Residual water0.1%
View more
2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only