Thermo Scientific Chemicals

Toluene, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 031755.AP
500 mL, Each
Thermo Scientific Chemicals

Toluene, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog number: 031755.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 031755.AP
also known as 031755-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-88-3
IUPAC Name
toluene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1
Form
Liquid
Impurity content
Substances darkened by sulfuric acid P.T.
Impurity content
Sulfuric compounds (as S); ≤0.003%
Water Content (Karl Fischer Titration)
≤0.03%
Residue on Evaporation
≤0.001%

Description

Toluene is used as solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text