Thermo Scientific Chemicals

N,N,N'-Trimethylethylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: 031785.06
5 g, Each
Thermo Scientific Chemicals

N,N,N'-Trimethylethylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: 031785.06
5 g, Each
Quantity
Catalog number: 031785.06
also known as 031785-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-25-6
IUPAC Name
[2-(dimethylazaniumyl)ethyl](methyl)azanium
Molecular Formula
C5H16N2
InChI Key
HVOYZOQVDYHUPF-UHFFFAOYSA-P
SMILES
C[NH2+]CC[NH+](C)C
Comment
Purchased in the U.S. and in other countries
Form
Liquid
Assay from Suppliers CofA
≥95.0% (GC, U.S. specification)
Refractive Index
1.4180-1.4210 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless to yellow

Description

N,N,N'-Trimethylethylenediamine is used as a precursor in the synthesis of a novel gemini surfactant viz. N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides. It is also used as an amine component for in-situ formation of alpha-amino alkoxides. Further, it is involved in the synthesis of N,N'-dimethyl-N'-(2-hydroxy-aethyl)-aethylendiamin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
  • Used as precursor in synthesis of novel gemini surfactant viz. N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides
  • Used as amine component for in-situ formation of alpha-amino alkoxides Involved in synthesis of N,N'-dimethyl-N'-(2-hydroxy-aethyl)-aethylendiamin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text