Thermo Scientific Chemicals

2,2,4-Trimethylpentane, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 031787.K7
4 L, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 031787.K7
4 L, Each
Quantity
Catalog number: 031787.K7
also known as 031787-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Appearance (Color)
Colorless
Color (APHA)
10, max
Assay (GC)
≥99.0%
Free acid (titration)
0.003meq/g, max (in the water soluble fraction)
Impurity content
Sulfur compounds (as S): 0.005%, max

Description

Solvent, thinner, in determination of fuel octane numbers2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as a thinner. It is involved as a processing aid in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text